missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ EZ-Run™ Protein Marker
Mixture of seven purified proteins optimized to produce uniform, compact bands when analyzed by SDS-PAGE and stained with Coomassie™ Blue
57.30 € - 126.00 €
Specifications
Product Type | Protein Molecular Weight Marker/Ladder |
---|---|
Form | Liquid |
Formulation | Supplied in gel-loading buffer |
For Use With (Application) | Monitoring protein separation during SDS-PAGE |
Kit Contents | <51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet |
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
10704914
|
Fisher BioReagents™
BP3600-500 |
500 μL, 100 Loadings | 100 Loadings |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10316323
|
Fisher BioReagents™
BP3600-1 |
2 x 500 μL, 200 Loadings | 200 Loadings |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Supplied in gel loading buffer for direct application to SDS-PAGE gels
- Ideal for molecular weight determination of polypeptides analyzed in various gel systems
Specifications
Protein Molecular Weight Marker/Ladder | |
Supplied in gel-loading buffer | |
<51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet | |
−20°C |
Liquid | |
Monitoring protein separation during SDS-PAGE | |
14.4 to 116 kd |