missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Lymphocyte Separation Medium-LSM
One-step centrifugation permits separation of mononuclear lymphocytes from erythrocytes, polynuclear lymphocytes and most platelets
Brand: Fisher BioReagents™ BP2663-500
Code : 32
Additional Details : Weight : 0.50000kg
Description
- Formulated for isolation of mononuclear cells from defribrinated or heparinized whole human blood
- Sterile-filtered solution of a sucrose polymer and diatrizoate salts at a specific gravity of 1.077 to 1.080g/mL at 20°C
Specifications
Animal Culture | |
Sterile | |
Separation of mononuclear lymphocytes from erythrocytes, polynuclear lymphocytes and most platelets |
Lymphocyte separation medium | |
Poly Bottles | |
5 x 100 mL |