missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Nucleotides
Nucleotides for PCR with a purity level of >98% triphosphate
92.00 € - 270.00 €
Specifications
Format | Liquid |
---|---|
pH | 7 to 7.5 |
Concentration | 100 mM |
Purity | >98% |
Color | Colorless |
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
10658973
|
Fisher BioReagents™
BP2560-4 |
dATP | 400 μL |
114.00 €
400µL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10410274
|
Fisher BioReagents™
BP2562-4 |
dCTP | 400 μL |
108.00 €
400µL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10152863
|
Fisher BioReagents™
BP2561-4 |
dGTP | 400 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10336653
|
Fisher BioReagents™
BP2564-1 |
dNTP Set | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10234683
|
Fisher BioReagents™
BP2564-4 |
dNTP Set | 400 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Functionally tested in PCR
- Can be used in dideoxy sequencing, mutagenesis, and cDNA synthesis applications
- DNase-, RNase-, and protease-free
- PCR dNTP Mix is a solution of dATP, dCTP, dGTP, and dTTP
- dNTP Set contains one tube each of dATP, dCTP, dGTP, and dTTP (4 tubes/set)
Specifications
Liquid | |
100 mM | |
Colorless |
7 to 7.5 | |
>98% | |
Dideoxy sequencing, mutagenesis and cDNA synthesis |