missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ NZY Broth (Powder/Molecular Genetics)

Brand:  Fisher BioReagents™ BP2465-10

Code : A0

Additional Details : Weight : 10.00000kg

 View more versions of this product

Product Code. 10002864

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

Keep refrigerated. (Store below 4°C/39°F.) Store in a cool, dry, well-ventilated area.
Molecular genetics
Microbiology Media
10 kg
Powder
NZY Broth
Poly Pail
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers