missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ Polymer Membrane Combination ISEs, Mercury-Free

Fast, precise measurements of calcium, nitrate and potassium ion in aqueous solutions over a wide range of concentrations

620.00 € - 665.00 €

Specifications

Diameter (Metric) Body 12.7mm
Diameter (Metric) Cap 15.8mm
Connector Types BNC
Material Epoxy Body
Length (Metric) 127mm
View More Specs

Products 3
Product Code Brand Filling Solution Type pH Range Resistance Electrode Price Quantity & Availability  
Product Code Brand Filling Solution Type pH Range Resistance Electrode Price Quantity & Availability  
11510144
View Documents
Fisherbrand™
4mol/L KCl with AgCl 2.5 to 11 10 to 20megohm Potassium 645.00 €
Each View alternative products
Save Up to 
»
Estimated Shipment
25-09-2020
Add to basket
 
11520144
View Documents
Fisherbrand™
0.1M (NH4)2SO4/0.01mol/L KCl with Ag+ 2 to 12 100megohm Nitrate 665.00 €
Each View alternative products
Save Up to 
»
Estimated Shipment
25-09-2020
Add to basket
 
11530144
View Documents
Fisherbrand™
4mol/L KCl with AgCl 3 to 10 >1megohm Calcium 620.00 €
Each View alternative products
Save Up to 
»
Estimated Shipment
25-09-2020
Add to basket
 
Description

Description

Sensor sealed in epoxy body

  • Sensing modules contain liquid internal filling solution in contact with gelled organophilic membrane
  • Membrane is impregnated with a selective ion exchanger for ion of interest
  • Sensors are sealed into durable epoxy body with built-in reference half-cell
  • Permit fast, precise measurements in aqueous solutions over a wide range of concentrations
  • Epoxy body provides chemical resistance
Features
  • Tough, chemical-resistant epoxy body
  • Membrane assembly sealed to body
  • Continuous electrical shielding and insulation of internal elements, cable and connectors
  • Extremely stable, reproducible readings with a minimum of electrical noise
  • Lower detection limits than conventional ISEs
  • Rapid response time that lowers cost per test
  • Individually tested, serialized to meet GLP requirements and backed by the Fisher Scientific Technical Applications staff
Dimensions
  • Diameter: body, 0.5 in. (12.7mm); cap, 0.625 in. (16mm)
  • Length: body, 4 in. (10.2cm); cap, 1 in. (2.5cm); overall, 5 in. (12.7cm)

Calcium Electrode

  • Finds application in the testing of wine, beer, dairy products, foods, plants, pharmaceuticals, fertilizers, explosives, clay and soil
  • Used as an endpoint detector in EDTA calcium titrations
  • Filling solution is 4M KCl (with AgCl)
Nitrate Electrode
  • Used in pollution testing, bioresearch, pharmaceutical production
  • Determines nitrate in soils, plants, sewage, fertilizers, explosives, meats, pickling baths
  • Filling solution is 0.1M (NH4)2SO4/0.01M KCl (with Ag+)
Potassium Electrode
  • Aids in analysis of serum, urine, saliva, other bodily fluids
  • Used in testing soils, sewage, fertilizers, foods and beverages
  • Filling solution is 5M NaCl

Specifications

Specifications

12.7mm
BNC
127mm
0° to 40°C
Cl2-, NO-, Br-, CN-, CIO-3, I-, CIO4-
15.8mm
Epoxy Body
∽1 min. or less
0 to 40°C