Learn More
Fisherbrand™ Circular Bottom Desi-Vac™ Container Desiccators with Vacuum Pump
Desiccator with built-in manual vacuum pump eliminates need for separate pump, tubing and stopcock
Brand:  Fisherbrand™
Code : 41
Additional Details : Weight : 0.29516kg
 View more versions of this product
Product Code. 11744366
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alert:
Do not autoclave or expose container to organic solvents or their vapors. Do not use concentrated acids as desiccants.
Description
Description
Desiccator with built-in manual vacuum pump eliminates need for separate pump, tubing and stopcock
  • Ideal for drying, preserving, storing and shipping samples
  • Self-contained unit holds a vacuum of 14.7 in.Hg (373mm) in excess of 24 hours
  • Rubber O-ring in cover ensures a tight seal
  • To remove cover, simply push release button to allow air into container
  • Clear polycarbonate body, metal pump handle, chemical-resistant ABS plastic pump and cover
  • No plate required
Ideal for drying, preserving, storing and shipping samples
Specifications
Specifications
Circular
Polycarbonate
Plastic pump handle, chemical-resistant ABS plastic vacuum pump and cover, release button on top, O-Ring-sealed cover
700 mL
11.43 cm
5 in.
12.7 cm
11.43 x 11.43 x 12.7cm ((4.5 x 4.5 x 5.0 in.)
Desiccator
42 cu. in.
4.5 in.
12.7 cm
5.0 in.