missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ MultiTube Vortexers: Centrifuge Tube Racks
Centrifuge tube racks for Fisherbrand™ MultiTube Vortexers.
34.25 € - 47.55 €
Specifications
Product Type | Centrifuge Tube Rack |
---|---|
For Use With (Equipment) | Multitube vortexer |
Product Code | Brand | No. of Positions | Holds | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Positions | Holds | Color | Price | Quantity & Availability | |||
15599004
|
Fisherbrand™
|
50 | 15 mL | Green |
47.55 €
Each |
In Stock
Log in to see stock. |
|||
15519014
|
Fisherbrand™
|
17 | 50 mL | Red |
34.25 €
Each |
In Stock
Log in to see stock. |
|||
Description
Centrifuge tube racks for Fisherbrand™ MultiTube Vortexers.
For use with Fisher Scientific MultiTube Vortexers
Color-coded foam racks for centrifuge tubes
Specifications
Centrifuge Tube Rack | |
Multitube vortexer |