missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Clamps for Owl P10DS Dual Gel System
Clamps for Owl&trade: P10DS Dual Gel System
Brand: Fisherbrand™
Code : 04
Additional Details : Weight : 0.22000kg
Description
- For use with Owl P10DS dual gel system
Specifications
Owl™ P10DS Dual Gel System | |
20 x 20 cm | |
20 cm | |
20 cm |
Side Clamp | |
7.8 in. | |
7.8 in. |