missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ FB-VE10-1 Vertical Electrophoresis System: Glass Plates

Notched or blank glass

Brand:  Fisherbrand™

Code : 04

Additional Details : Weight : 0.04800kg

 View more versions of this product

Product Code. 11963065

  • 23.90 € / Each
In Stock
to see stock.
Add to basket

Description

Description

Specifications

Specifications

FisherBiotech FB-VE10-1 Vertical Electrophoresis System; Owl™ P7-CST Multiple Gradient Gel Caster
Glass
0.24 cm
10 cm
0.24 cm
Blank Glass Plate
0.09 in.
10 cm
0.09 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers