missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Horizontal Large Gel Systems: Gels

For use with large gel systems

Brand:  Fisherbrand™

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11962925

  • 195.00 € / Each


Estimated Shipment
05-08-2021
Add to basket

Description

Description

UV-transparent; allows multiple gels to be poured.

Specifications

Specifications

UV Transparent
Owl™ A2 and A5 Large Gel Electrophoresis System
4.31 in.
Gasketed end gate
14.75 in.
37.5 cm
11 in.
28 cm
9.8 x 7.8 in. (25 x 20 cm)
Gel Tray
11 cm
9.8 in.
25 cm
7.8 in.
20 cm
Videos
SDS
Documents