missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Spacers for Owl™ P10DS Dual Gel System
Spacers for Owl™ P10DS Dual Gel System
Brand: Fisherbrand™
Code : 04
Additional Details : Weight : 0.00800kg
Description
- For use with Owl P10DS dual gel system
Specifications
Owl™ P10DS Dual Gel System | |
20 x 20 cm | |
9.05 in. | |
23 cm | |
7.8 in. | |
20 cm | |
0.08 cm | |
7.8 in. | |
13 cm |
Spacer Set | |
0.03 in. | |
0.08 cm | |
9.84 in. | |
25 cm | |
0.03 in. | |
5.11 in. | |
0.1 cm | |
20 cm |