missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Absorption Quartz Micro Rectangular Cells with PTFE Lid
Fit standard spectrophotometer cell holders
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Fit standard spectrophotometer cell holders
- All cells are 12.5mm wide
- 10mm path length rectangular cells
- Reduce reagent use with Micro cells
- Quartz windows for transmission from far ultraviolet through the infrared
Specifications
Micro Absorption Cell | |
Quartz | |
2mm | |
2/Pk., 8 Pk./Cs. | |
200 to 2500 nm | |
Standard Spectrophotometer Cell Holder | |
Transmission from far ultraviolet through the infrared |
500 μL | |
PTFE | |
2 clear windows | |
10 mm | |
45 mm | |
12.5 mm | |
Rectangle |