missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Aerosol-Barrier Tips
Prevent cross-contamination by using superior aerosol-barrier tips.
77.50 €
Specifications
Filtered | Filtered |
---|---|
Color | Natural |
Clean Claims | DNase, RNase, DNA, and Pyrogen Free |
For Use With (Equipment) | Universal Fit Pipettor |
Product Code | Brand | Volume (Metric) | Autoclavable | Format | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Autoclavable | Format | Price | Quantity & Availability | |||
11917734
|
Fisherbrand™
|
0.1 to 5 μL | Not Autoclavable | Rack |
77.50 €
Pack of 960 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
Description
Fisherbrand™ Aerosol-Barrier tips can be used universally and protects the intergrity of your samples.
- Unique small micron filter prevents aerosol contamination with no obstruction to airflow
- Use aerosol barrier tips to ensure result integrity
- Certified by independent laboratory RNase, DNase, DNA, and pyrogen free
- Natural color
Packaging Code -- BP: Bulk Pack; RI: Rack Inserts; HR: Hinged Rack; HRS: Hinged Rack, Sterile; HR/T: Hinged Rack, Trace Metal-free; LR: Lift-off Rack; LRS: Lift-off Rack, Sterile; SR: Stack Rack; IWS: Individually Wrapped, Sterile.
Specifications
Filtered | |
Natural | |
DNase, RNase, DNA, and Pyrogen Free | |
Universal Fit Pipettor |