missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable Bottletop Dispensers
Provides smooth, effortless dispensing
242.00 € - 335.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Includes | 13-in. inlet tube (with extension) |
Includes: Calibration certificate
Product Code | Brand | Accuracy | Increments | Precision | Volume (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Accuracy | Increments | Precision | Volume (Metric) | Price | Quantity & Availability | ||
12867913
|
Fisherbrand™
|
±0.6% (±0.015mL) | 0.05 mL | ±0.2% (±0.005mL) | 0.25 to 2.5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12877913
|
Fisherbrand™
|
±0.6% (±0.030mL) | 0.1 mL | ±0.2% (±0.010mL) | 0.5 to 5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12897913
|
Fisherbrand™
|
±0.6% (±0.180mL) | 0.5 mL | ±0.2% (±0.060mL) | 2.5 to 30 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12807923
|
Fisherbrand™
|
±0.6% (±0.360mL) | 1.0 mL | ±0.2% (±0.120mL) | 5 to 60 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12817923
|
Fisherbrand™
|
±0.6% (±3.000mL) | 2.0 mL | ±0.2% (±0.200mL) | 10 to 100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Provides smooth, effortless dispensing
- Wetted parts are PTFE, FEP and borosilicate glass (allows instrument to be used with a wide variety of chemicals)
- 7 volume ranges to choose from
- Bubble-free dispensing
- Effortless plunger movement
- Disassembles for easy cleaning and servicing
- Autoclavable at 121°C, 15 psi for 10 to 15 minutes
- Each bottle top dispenser includes one 28mm, 32mm, 36mm, 40mm, 45mm bottle neck adaptor
Component Description
- Piston: PTFE
- Cylinder: Glass
- Volume Adjustment Knob: Screw Type
- Valve Housing: PTFE
- Valve Assembly: Glass Ball & Seal
- Discharge Assembly: PTFE
- Delivery Tube: FEP
- Inlet Tube: FEP, 13 in. (with extension)
- Calibration: Individually calibrated and certified (in-lab by user is also possible)
- Accuracy & Reproducibility: In accordance with ISO 8655 standards
- Compability: Excellent with most reagents
Certifications
ISO 8655
Warranty
One year free replacement warranty against any manufacturing defects. The period of one year starts from the invoice date of Fisher Scientific to its customer.
Specifications
Autoclavable | |
13-in. inlet tube (with extension) |