missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cork Stoppers - Superior Grade

Corks are cut from high-quality wood and are selected to meet superior standards

Brand:  Fisherbrand™

Additional Details : Weight : 0.12500kg

 View more versions of this product

Product Code. 16466520

  • 27.00 € / Pack of 500
Estimated Shipment: 26-06-2024
to see stock.
Add to basket

Description

Description

Corks are cut from high-quality wood and are selected to meet superior standards

Specifications

Specifications

Natural
0
0
0.96 cm
Wood
1.27 cm
Cork Stopper
Ideal for sealing bottles
0.71 cm
500/Pk.
Superior
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers