Learn More
Fisherbrand™ Fisherbrand™ Large-Orifice Pipet Tips
Fisherbrand™ Large-Orifice Pipet Tips helps you pipet difficult liquids.
POA
Specifications
Tip Style Genomic/Wide Orifice
View More Specs
Description
Description
Genomic/Wide Orifice assists in pipetting small volumes of viscous materials.
  • Assist in pipetting small volumes of viscous materials
  • Help eliminate shearing of genomic DNA and fragile cells
Specifications
Specifications
Genomic/Wide Orifice