missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable Biohazard Waste Bags
Printed in English and Spanish
86.98 € - 261.76 €
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | ASTM D1709, Dart impact |
Closure Type | Wire Tie |
Color | Orange |
Material | Polypropylene |
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
18326533
|
Fisherbrand™
|
256.08 €
Pack of 100 |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18396503
|
Fisherbrand™
|
86.98 €
Pack of 200 |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18326513
|
Fisherbrand™
|
261.76 €
Pack of 200 |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Printed in English and Spanish
- Max. temperature of 140°C (285°F); use at 125°C (257°F)
- Come with wire-tie closures
- Constructed of polypropylene
Specifications
Autoclavable | |
Wire Tie | |
Polypropylene | |
140°C |
ASTM D1709, Dart impact | |
Orange | |
285°F |