Learn More
Fisherbrand™ Suprasil™ Quartz Semi-Micro Cell Cuvettes
Designed to hold samples for spectroscopic experiments and optical analysis. Fisherbrand™ Suprasil™ Quartz Semi-Micro Cell Cuvettes are supplied with either a PTFE cover or plug.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Sold in pairs
- Very chemically resistant
- Maintains shape, even at high temperatures up to approximately 1000°C