missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Silicone Bulb-Type Safety Pipet Fillers

Chemical-resistant, resilient, silicone pipet fillers with glass valves improve worker safety

Brand:  Fisherbrand™

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 15936522

  • 52.00 € / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Improve worker safety with these chemical-resistant, resilient, silicone pipet fillers with glass valves.

  • Glass valves improve worker safety by eliminating mouth pipetting and handling of hazardous materials
  • Long-lasting silicone is highly resistant to organic solvents
  • Fit any pipet with O.D. ranging from 0.19 to 0.38 in. (4.5 to 9 mm)

To Use:

  • Fill and dispense pipets with one hand
  • Expel air by squeezing valve '1' above bulb
  • Draw up liquid by pressing valve '2' on stem
  • To release, press valve '3' on side of stem
Specifications

Specifications

No
Not Autoclavable
Pipette
3 Valve Standard
Pipette Filler
Pipet
4.5 mm to 9 mm
Red
Silicone
1
50 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers