missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Single Cytology Funnels
For the Epredia™ Shandon™ CytoSpin™ Centrifuge
5189.90 €
Product Code | Brand | Color | Quantity | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Quantity | Volume (Metric) | Price | Quantity & Availability | |||
15201886
|
Fisherbrand™
|
White | 500/Cs. | Up to 0.4 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
For the Thermo Scientific™ Shandon™ CytoSpin™ Centrifuge
- Functional equivalent of the Shandon Single Cytofunnel™
- Fully compatible with Shandon CytoSpin Centrifuge
- White Filter Card is pre-attached and is designed for single-use only
- Brown Filter Card for slower absorption of sparse samples — ideal for CSF
- For sample volumes of up to 0.4mL or less
- Provides consistent and homogeneous cell deposition area of 6mm
Specifications
Shandon CytoSpin Centrifuge |