Learn More
Motic™ FITC Filter Block, BA310E
For detection of FITC fluorophore. Motic™ FITC Filter Block, BA310E is suitable for BA310E Microscopes.
Brand:  Motic™ 1101001902923
Additional Details : Weight : 0.01000kg
Product Code. 15495242
  • 2000.00 € / Each
Estimated Shipment: 04-07-2024
to see stock.
Add to basket
Description
Description
  • Epi-LED fluorescence equipment for FITC (blue) excitation.
  • Block houses an excitation filter, an emission filter, and a dichroic beam splitter.
  • Suitable for BA-310E Microscopes.
Specifications
Specifications
Filter Block, FITC
BA-310E Microscopes
Excitation Filter, Emission Filter, Dichroic Beam splitter