Learn More
Flat-bottom Glass Tube with Screw Thread
Free-standing Duran™ Flat-bottom Glass Tube with Screw Thread.
31.95 € - 37.20 €
Specifications
Product Type Flat Bottom Tube
Quantity 1Each
View More Specs
Products 2
Product Code Brand Diameter (Metric) Outer Length (Metric) Thread Size Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Length (Metric) Thread Size Price Quantity & Availability  
11752919 Duran™
248370105
12mm 100mm GL14 DIN Thread 37.20 €
Pack of 10
Estimated Shipment
13-02-2019
Add to basket
11742919 Duran™
248360207
12mm 100mm GL14 DIN Thread 31.95 €
Pack of 10
Estimated Shipment
13-02-2019
Add to basket
Description
Description
  • Durable Duran™ glass construction
  • DIN thread (GL 14)
  • 10/Pk
Specifications
Specifications
Flat Bottom Tube
1Each