missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flt-3 Inhibitor, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mg
50 mg
Description
A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor
Specifications
Quantity | 50 mg |
Description | White |
Content And Storage | 2°C to 8°C |
For Use With (Application) | A cell-permeable thienylcarboxamide compound that acts as a potent, ATP-competitive, and highly selective Flt-3 inhibitor |
Formula Weight | 360.43g/mol |
Sensitivity | Light Sensitive |
Product Type | Signal Transduction Reagent |
Form | Powder |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'