missing translation for 'onlineSavingsMsg'
Learn More

Flubendazole, Thermo Scientific Chemicals

Catalog Number p-7077141
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
31430-15-6
C16H12FN3O3
313.28
CPEUVMUXAHMANV-UHFFFAOYSA-N
flubendazole, flumoxane, flubendazol, fluvermal, flubenol, flubendazolum, flumoxal, flutelmium, methyl 5-4-fluorobenzoyl-1h-benzo d imidazol-2-yl carbamate, flubendazol inn-spanish
35802
CHEBI:77095
methyl N-[6-(4-fluorobenzoyl)-1H-benzimidazol-2-yl]carbamate
COC(=O)NC1=NC2=C(N1)C=C(C=C2)C(=O)C3=CC=C(C=C3)F
This item is not returnable. View return policy
31430-15-6
C16H12FN3O3
313.28
CPEUVMUXAHMANV-UHFFFAOYSA-N
flubendazole, flumoxane, flubendazol, fluvermal, flubenol, flubendazolum, flumoxal, flutelmium, methyl 5-4-fluorobenzoyl-1h-benzo d imidazol-2-yl carbamate, flubendazol inn-spanish
35802
CHEBI:77095
methyl N-[6-(4-fluorobenzoyl)-1H-benzimidazol-2-yl]carbamate
COC(=O)NC1=NC2=C(N1)C=C(C=C2)C(=O)C3=CC=C(C=C3)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 260.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 4 hr)
Infrared Spectrum Authentic
Quantity 1 g
Formula Weight 313.28
Chemical Name or Material Flubendazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'