Learn More
Thermo Scientific™ Fluid Transfer Assembly, Extension Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
236.00 € - 2140.00 €
Product Code | Brand | Tubing Style | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Style | Price | Quantity & Availability | |||||
17850687
|
Thermo Scientific™
SH31116.01 |
9.52 mm (3/8 in.) ID; 15.88 mm (5/8 in.) OD; TPE 374; 1 m (3.25 ft); MPC |
1215.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17880687
|
Thermo Scientific™
SH31116.02 |
9.52 mm (3/8 in.) ID; 15.88 mm (5/8 in.) OD; TPE 374; 2.5 m (8.2 ft); MPC |
491.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17860637
|
Thermo Scientific™
SH31116.03 |
9.52 mm (3/8 in.) ID; 15.88 mm (5/8 in.) OD; TPE 374; 5 m (16.4 ft); MPC |
1970.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17810697
|
Thermo Scientific™
SH31117.01 |
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 1 m (3.25 ft); Luer |
1045.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17890687
|
Thermo Scientific™
SH31117.02 |
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 2.5 m (8.2 ft); Luer |
468.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17800697
|
Thermo Scientific™
SH31117.03 |
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 5 m (16.4 ft); Luer |
1845.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17820697
|
Thermo Scientific™
SH31118.01 |
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 1 m (3.25 ft); MPC |
253.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17830647
|
Thermo Scientific™
SH31118.02 |
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 2.5 m (8.2 ft); MPC |
236.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17800647
|
Thermo Scientific™
SH31118.03 |
6.35 mm (1/4 in.) ID; 11.11 mm (7/16 in.) OD; TPE 374; 5 m (16.4 ft); MPC |
730.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17890567
|
Thermo Scientific™
SH31119.01 |
3.22 mm (1/8 in.) ID; 6.35 mm (1/4 in.) OD; TPE 374; 1 m (3.25 ft); Luer |
1020.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17820647
|
Thermo Scientific™
SH31119.02 |
3.22 mm (1/8 in.) ID; 6.35 mm (1/4 in.) OD; TPE 374; 2.5 m (8.2 ft); Luer |
412.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17840647
|
Thermo Scientific™
SH31119.03 |
3.22 mm (1/8 in.) ID; 6.35 mm (1/4 in.) OD; TPE 374; 5 m (16.4 ft); Luer |
1720.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17850647
|
Thermo Scientific™
SH31120.01 |
12.7 mm (1/2 in.) ID; 19.05 mm (3/4 in.) OD; TPE 374; 1 m (3.25 ft); MPX |
1545.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17860647
|
Thermo Scientific™
SH31120.02 |
12.7 mm (1/2 in.) ID; 19.05 mm (3/4 in.) OD; TPE 374; 2.5 m (8.2 ft); MPX |
840.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17810577
|
Thermo Scientific™
SH31120.03 |
12.7 mm (1/2 in.) ID; 19.05 mm (3/4 in.) OD; TPE 374; 5 m (16.4 ft); MPX |
2140.00 €
Pack of 10 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Extension Set |