missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMC63 scFv Mouse, FITC, Clone: Y45, AcroBiosystems
Mouse Monoclonal Antibody
327.00 € - 2020.00 €
Specifications
| Antigen | FMC63 scFv |
|---|---|
| Clone | Y45 |
| Classification | Monoclonal |
| Conjugate | FITC |
| Host Species | Hybridoma |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16851936
|
ACROBiosystems
FM3-FY45-25tests |
25 Tests |
327.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16841936
|
ACROBiosystems
FM3-FY45-200tests |
200 Tests |
2020.00 €
200 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FMC63 scFv Monoclonal antibody specifically detects FMC63 scFv.Specifications
| FMC63 scFv | |
| Monoclonal | |
| Hybridoma | |
| IgG1 | |
| -20°C, aviod light |
| Y45 | |
| FITC | |
| RUO | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title