missing translation for 'onlineSavingsMsg'
Learn More

FMOC-D-phenylalanine, 98%, Thermo Scientific Chemicals

Catalog Number p-4323581
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
86123-10-6
C24H21NO4
387.44
MFCD00062955
SJVFAHZPLIXNDH-JOCHJYFZSA-N
fmoc-d-phenylalanine, fmoc-d-phe-oh, n-fmoc-d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-beta-phenylalanine, n-9-fluorenylmethoxycarbonyl-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylalanine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-phenyl-propanoic acid
978332
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
OC(=O)[C@@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
This item is not returnable. View return policy
86123-10-6
C24H21NO4
387.44
MFCD00062955
SJVFAHZPLIXNDH-JOCHJYFZSA-N
fmoc-d-phenylalanine, fmoc-d-phe-oh, n-fmoc-d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-beta-phenylalanine, n-9-fluorenylmethoxycarbonyl-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylalanine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-phenyl-propanoic acid
978332
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
OC(=O)[C@@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 184.0°C to 186.5°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation + 40.00
Formula Weight 387.43
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material FMOC-D-phenylalanine
Show More Show Less
compliance-icons
Product Identifier
  • FMOC-D-phenylalanine, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'