missing translation for 'onlineSavingsMsg'
Learn More

FMOC isothiocyanate, 97%, ACROS Organics™

Brand:  Acros Organics 438660010

Code : 29

Additional Details : CAS Number : 199915-38-3 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11945761

  • 95.34 € / 1g

Estimated Shipment: 08-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

199915-38-3
281.34
fmoc isothiocyanate, fmoc-isothiocyanate, 9-fluorenylmethoxycarbonyl isothiocyanate, 9h-fluoren-9-ylmethyl n-sulfanylidenemethylidene carbamate, 9h-fluoren-9-ylmethyl n-carbothioylcarbamate, n-fmoc-isothiocyanate, 9-fluorenylmethyl isothiocyanatoformate, 9-fluorenylmethoxycarbonylisothiocyanate, fmoc isothiocyanate chn, fluoren-9-ylmethyloxycarbonyl isothiocyanate
9H-fluoren-9-ylmethyl N-(sulfanylidenemethylidene)carbamate
C16H11NO2S
DHMYULZVFHHEHE-UHFFFAOYSA-N
4154759
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)N=C=S

Specifications

FMOC isothiocyanate
Authentic
Yellow to Brown
1g
97%
fmoc isothiocyanate, fmoc-isothiocyanate, 9-fluorenylmethoxycarbonyl isothiocyanate, 9h-fluoren-9-ylmethyl n-sulfanylidenemethylidene carbamate, 9h-fluoren-9-ylmethyl n-carbothioylcarbamate, n-fmoc-isothiocyanate, 9-fluorenylmethyl isothiocyanatoformate, 9-fluorenylmethoxycarbonylisothiocyanate, fmoc isothiocyanate chn, fluoren-9-ylmethyloxycarbonyl isothiocyanate
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)N=C=S
281.34
281.34
97%
65.6 to 71.0 % (C) 4.8 to 5.2 % (N)
Glass bottle
44.0°C to 45.0°C
199915-38-3
C16H11NO2S
DHMYULZVFHHEHE-UHFFFAOYSA-N
9H-fluoren-9-ylmethyl N-(sulfanylidenemethylidene)carbamate
4154759
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • FMOC isothiocyanate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS Documents
Certificates