Learn More
FMOC-L-arginine(PMC), 95%, ACROS Organics™
Brand: Acros Organics 439870010
Code : 29
Additional Details : CAS Number : 119831-72-0 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
119831-72-0 | |
662.81 | |
fmoc-arg pmc-oh, o-2,2,5,7,8-pentamethylchromane-6-sulfonyl-l-arginine, fmoc-l-arg pmc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,5,7,8-pentamethylchroman-6-yl sulfonyl guanidino pentanoic acid, fmoc-arg pmc, fmoc-arg pmc oh, ambotzfaa1351, n,a-fmoc-n, o-pmc-l-arginine, n-,a-fmoc-n-, o-2,2,5,7,8-pentamethylchroman | |
(2S)-5-[[amino-[(2,2,5,7,8-pentamethyl-3,4-dihydrochromen-6-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid |
C35H42N4O7S | |
QTWZCODKTSUZJN-LJAQVGFWSA-N | |
3010429 | |
CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CCC(O2)(C)C |
Specifications
(1mmol in 1mL DMF) clear, colorless | |
Authentic | |
-4.5 | |
108°C to 109°C | |
119831-72-0 | |
C35H42N4O7S | |
QTWZCODKTSUZJN-LJAQVGFWSA-N | |
(2S)-5-[[amino-[(2,2,5,7,8-pentamethyl-3,4-dihydrochromen-6-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid | |
3010429 | |
Powder |
FMOC-L-Arginine(PMC) | |
Glass Bottle | |
White | |
1g | |
94% min. (HPLC) | |
fmoc-arg pmc-oh, o-2,2,5,7,8-pentamethylchromane-6-sulfonyl-l-arginine, fmoc-l-arg pmc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,5,7,8-pentamethylchroman-6-yl sulfonyl guanidino pentanoic acid, fmoc-arg pmc, fmoc-arg pmc oh, ambotzfaa1351, n,a-fmoc-n, o-pmc-l-arginine, n-,a-fmoc-n-, o-2,2,5,7,8-pentamethylchroman | |
CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)N)C)CCC(O2)(C)C | |
662.81 | |
662.81 | |
95% |
Safety and Handling
- Fmoc-L-Arginine(Pmc)
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.