missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCHOTT AG Lighting and Imaging™ Focusing Lens with Filter set
For use with flexible or gooseneck light guides up to 5mm diameter. Includes red, blue, green, and yellow filter set.
Brand: SCHOTT AG Lighting and Imaging™ 158200
This item is not returnable.
View return policy
Description
- Fits flexible or gooseneck lightguides with a diameter up to 5mm
- Includes red, blue, green, yellow filter set
- Light intensity at the spot center can be increased by factor 3.
Specifications
| Lens and Filter Set | |
| Stereo Microscopy Illumination | |
| Filter set: red, blue, green, yellow |
| 5 mm | |
| Flexible or Gooseneck Light Guides |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction