missing translation for 'onlineSavingsMsg'
Learn More

Edulab™ Force board wall type 750mm x 600mm

Brand:  Edulab™ P348

Code : 50

Additional Details : Weight : 2.00000kg

Product Code. 13181991

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.