missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde, Technical, Solution, Stabilised In Methanol, Fisher Chemical™
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
Specifications
Packaging | Plastic container |
Quantity | 25 L |
Color | Colorless |
Formula Weight | 30.02g/mol |
Vapor Pressure | 2mbar at 20°C |
Vapor Density | > 1.0 |
pH | 3 to 4.2 |
Melting Point | -15°C |
Boiling Point | 97°C |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Formaldehyde solution, 37%
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H370-Causes damage to organs.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Formaldehyde, Methylalcohol
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'