missing translation for 'onlineSavingsMsg'
Learn More

Formamide, Ultra Pure, 99.5% min, Thermo Scientific Chemicals

Catalog Number p-7057714
Click to view available options
Quantity:
100 mL
250 mL
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O

Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.

Solubility
Soluble in water (20 mg/ml ), methanol, ethanol and acetone. Slightly soluble in ether.

Notes
Hygroscopic. Store at room temperature. Store away from water or moisture.
TRUSTED_SUSTAINABILITY
Melting Point 2°C
Color Colorless
Density 1.134 g/mL
Boiling Point 210°C
Quantity 100 mL
Flash Point 150°C (302°F)
Odor Ammonia-like
Physical Form Liquid
Refractive Index 1.4475
Beilstein 505995
Solubility Information Soluble in water (20mg/ml ),methanol,ethanol and acetone. Slightly soluble in ether.
Formula Weight 45.04
Percent Purity ≥99.5%
Chemical Name or Material Formamide, Ultra Pure
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'