missing translation for 'onlineSavingsMsg'
Learn More

Formamide, Ultrapure Bioreagent, J.T. Baker™

Catalog Number p-5383260
Click to view available options
Quantity:
100 mL
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
This item is not returnable. View return policy
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For denaturing nucleic acids prior to analysis
Formula Weight 45.04
Grade Ultra Pure
Quantity 100 mL
Flash Point 154.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.13kg
Chemical Name or Material Formamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'