missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid, 98-100%, Certified AR for Analysis, Fisher Chemical™
27.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Plastic Coated Glass bottle
Plastic container
Description
CH2O2, CAS Number-64-18-6, formylic acid, myrmicyl, acide formique, methanoic acid, hydrogen carboxylic acid, aminic acid, formira, bilorin, collo-bueglatt, formisoton, 2.5L, 101 deg.C, CHEBI:30751, Colorless, 46.02g/mol, BDAGIHXWWSANSR-UHFFFAOYSA-N, formic acid, 8 deg.C, 3297, 46.025
Specifications
pH | 2.1 |
Melting Point | 8°C |
Boiling Point | 101°C |
Color | Colorless |
Formula Weight | 46.02g/mol |
Quantity | 500 mL |
Packaging | Plastic Coated Glass bottle |
Physical Form | Liquid |
Vapor Pressure | 44mbar at 20°C |
Safety and Handling
Product Identifier
- Formic acid
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- EUH071-Corrosive to the respiratory tract.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'