missing translation for 'onlineSavingsMsg'
Learn More

Formic acid, 99%, for analysis, Thermo Scientific Chemicals

Catalog Number p-3734368
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Plastic Bottle
Plastic drum
64-18-6
CH2O2
46.025
MFCD00003297
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O
64-18-6
CH2O2
46.025
MFCD00003297
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O

Formic acid, 99%, CH2O2, CAS Number-64-18-6, acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl, 2.5L, Vapor Pressure: 44mbar at 20 deg.C, 02, 8, 101.0 deg.C, 0.1ppm max., CHEBI:30751, 5ppm max., Undesignated

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY
Melting Point 8.0°C
Boiling Point 101.0°C
Assay Percent Range 99%
Linear Formula HCO2H
Beilstein 02, 8
Fieser 01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177
Merck Index 15, 4269
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol, ether and glycerine
Formula Weight 46.02
Percent Purity 99%
Grade Analytical
Density 1.2200g/mL
Quantity 25 mL
Flash Point 50°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Physical Form Liquid
Refractive Index 1.3700 to 1.3720
Specific Gravity 1.22
Vapor Pressure 44mbar at 20°C
Chemical Name or Material Formic acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'