missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
62.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.
36.87 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
62.78 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
36.87 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
62.78 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
0.5 mL
1 mL
10 Ampules
2 mL
50 mL
Packaging:
Ampule
Ampules
Nalgene™ Poly Bottle
Specifications
Absorbance | 0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm |
pH | 2.1 |
Melting Point | 8°C |
Boiling Point | 101°C |
Color | Colorless |
Assay Percent Range | ≥99.0 % |
Linear Formula | HCOOH |
Merck Index | 15, 4269 |
SMILES | C(=O)O |
Molecular Weight (g/mol) | 46.025 |
Show More |
Safety and Handling
Product Identifier
- Formic acid, OPTIMA LC/MS Grade (99.5%)
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- EUH071-Corrosive to the respiratory tract.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'