missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formic Acid Solution, Honeywell Fluka™
56.03 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Use promo code "21664" to get your promotional price.
Click to view available options
Quantity:
100 mL
500 mL
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Linear Formula | HCOOH |
UN Number | 3412 |
Beilstein | 1209246 |
Concentration | 49 to 51% (T) |
Formula Weight | 46.03g/mol |
Grade | puriss. p.a. |
Quantity | 100 mL |
Chemical Name or Material | Formic Acid Solution |
Safety and Handling
Product Identifier
- formic acid solution
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- EUH071-Corrosive to the respiratory tract.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : formic acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'