Learn More
Sartorius™ Formulation VI Q-App for Cubis™ MSA Balances
Customize a Cubis MSA scale to suit specific laboratory processes. Sartorius™ Formulation VI Q-App for Cubis™ MSA Balances is a standard application that delivers a simple formulation with integrated recipe memory.
Brand: Sartorius™ YAPP05
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Customize a Cubis MSA laboratory balance at any time by downloading specific applications, called Q-Apps, directly into the balance.
- Once installed, Q-Apps guide users step-by-step through a specific weighing workflow
- Integrating SOPs directly into the Cubis adapts the balance to a process
- Functions as embedded software on which fewer validation requirements are needed
- YAPPOF: Formulation VI – a simple formulation with integrated recipe memory
Specifications
Cubis™ MSA Series Balance | |
Simple formulation with integrated recipe memory | |
Cubis™ Q-Apps | |
Q-App License |