missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXA1 / HNF3A Mouse, Clone: FOXA1/1512, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3169-MSM2-P1
Additional Details : Weight : 0.01000kg
Specifications
FOXA1 / HNF3A | |
Nuclear | |
FOXA1/1512 | |
Unconjugated | |
NP_004487 | |
Mouse | |
79kDa | |
Protein A/G Purified | |
RUO | |
3169 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
FOXA1 | |
Recombinant human FOXA1 protein fragment (aa467-614) | |
HepG-2 or MCF-7 cells. Liver, Pancreas, Bladder, Prostate, Colon and Lung and Breast. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |