missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Foxp3 Mouse, PE-Cy7, Clone: 3G3, enQuireBio™
Mouse Monoclonal Antibody
360.00 € - 505.00 €
Specifications
Antigen | Foxp3 |
---|---|
Clone | 3G3 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932408
|
EnquireBio
QAB71-PE7-25UG |
25 μg |
360.00 €
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922408
|
EnquireBio
QAB71-PE7-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Foxp3 | |
0.2mg/mL | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.05% BSA, 0.05% NaN3, pH7.2 | |
20371 | |
IgG1 κ | |
Protein A/G Purified |
3G3 | |
Flow Cytometry | |
PE-Cyanine7 | |
RUO | |
NP_001186276 | |
Foxp3 | |
Primary | |
Store at 2-4°C. |