missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Frosted Microscope Slides, Cut

Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.

Brand:  Thermo Scientific™ AAAC0360076012EB0MNZ#0#G#

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15396518

  • 313.07 € / Pack of 100


Call for latest local availability
01 885 5854
Add to basket

Description

Description

  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
Specifications

Specifications

76mm
1mm
Uncoated
Soda-lime glass
Yes
36mm
N/A
20mm
100 pieces in paper
76 x 36mm
Videos