Learn More
Thermo Scientific™ Frosted Microscope Slides, Cut
Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.
23.80 € - 565.00 €
Product Code | Brand | Dimensions (L x W) | Frosted | Quantity | Writing Surface | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Frosted | Quantity | Writing Surface | Price | Quantity & Availability | ||
12392138
|
Thermo Scientific™
AAAA000001##13E01FST20PH# |
76 x 26 mm | 20 mm | 50/Box, 2000 in outer carton OEM | Yes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10782887
|
Thermo Scientific™
AAAA000001##02F00MNZ50MG# |
76 x 26 mm | Fully Frosted | 50/Box, 1000 in outer carton | No |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15363112
|
Thermo Scientific™
AAAC0500075012EB0MNZ#0#G# |
75 x 50 mm | 20 mm | 100 Pieces in Paper | Yes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Manufactured from Swiss extra-white soda-lime glass
- Reduced iron content for low self-fluorescence
- Greenish shimmer minimized
- Guaranteed optimum, faultless transparency
- Clean, flat and suitable for immediate use
Specifications
Soda Lime Glass | |
1 mm | |
Uncoated |