missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSH-Receptor Mouse, Clone: FSHR/1400, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2492-MSM1-P1
Additional Details : Weight : 0.01000kg
Specifications
FSH-Receptor | |
Cytoplasmic and Cell Surface | |
FSHR/1400 | |
Unconjugated | |
XP_011531035 | |
Mouse | |
75kDa | |
Protein A/G Purified | |
RUO | |
2492 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
FSHR | |
Recombinant human full-length FSHR protein | |
Uterine Carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |