missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 200 - Advanced, Programmable, Digital Heated Circulating Bath and Stainless Steel Tank

Provides outstanding performance for applications requiring accurate temperature control up to a max of 200°C. The heated, circulating bath includes a powerful integral pump for external applications. Can be paired with an immersion cooler for temperatures below ambient to a minimum of -15°C.

Brand:  Fisherbrand™ 17285576

 View more versions of this product

Product Code. 17285576

  • 3285.00 € / Each
In Stock
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Full color QVGA TFT screen clearly displays actual and set temperatures, pump speed and clear status icons
  • Optional external probes for monitoring and controlling temperature of remote loads
  • Intuitive screen icons and menus allow fast and accurate setup
  • USB and RS232 interface allows connection to PC or laptop for programming or data logging
  • Easy to configure automated temperature profiling and control functions
  • Can store up to 10 programs containing 100 segments
  • Powerful integral pump with multi-stage variable flow rate, maximum flow 22 L/min.
  • 5-point user calibration for optimum accuracy
  • Adjustable over-temperature cut-out
  • Variable high and low temperature alarm settings with configurable to switch a relay
  • 5 language options
  • PC software for program set up, data logging and real time graphing
  • Maximum cooling and heating range (dependent on tank size): -15° to +200°C with stainless steel tank
  • Stability: ± 0.01°C
  • Other accessories including lids, racks and immersion coolers sold separately
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 gal.
Air-cooled
22 L/min.
Heating Circulator with Open Bath
Stainless Steel
3 Years
220/240 V
38 L
-15°C (with immersion cooler)
5°C to 200°C
38 L Stainless Steel Tank
530 mbar
733 x 338 x 260 mm
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.