Learn More
Bochem™ Funnels
Made from 18/10 stainless steel. The Bochem Funnels are used to work with powders.
21.70 € - 208.00 €
Product Code | Brand | Height (Metric) | Diameter (Metric) Stem | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Diameter (Metric) Stem | Price | Quantity & Availability | ||||
10383811
|
Bochem™
8840 |
115 mm | 17/13mm |
21.70 €
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
10078550
|
Bochem™
8720 |
135 mm | 20mm |
208.00 €
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
Description
- Special funnel
- For powder
- Straight tube
- Made from 18/10 stainless steel
Specifications
Steel |