missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furegrelate Sodium Salt, ≥99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Description
Inhibitor of thrombaxane synthetase
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 50 mg |
Description | White to Off-white |
Content And Storage | -30°C to -10°C |
For Use With (Application) | Inhibitor of thrombaxane synthetase |
Formula Weight | 275.20g/mol |
Solubility | Soluble in water at approximately 15mg/ml |
Product Type | Signal Transduction Reagent |
Form | Crystalline solid |
Assay | ≥99% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'