Learn More
Carbolite™ Furnace with Type R Temperature Sensor
Brand:  Carbolite™ RHF1508-230SN+&02-301
 View more versions of this product
Product Code. 10009482
  • ONLINE EXCLUSIVE PRICE
    17365.00 €
    16562.28 € / Each
    Save 802.72 €
    4% Off
Only null left
Estimated Shipment: 02-01-2026
Add to basket
Specifications
Compliance to BSEN 61010
8 L
Furnace
270 x 170 x 170 mm
170 mm
170 mm
270 mm
230 V
50/60 Hz
301 PID Controller
1500°C
For use in quality control, heat treatment and research of ceramic materials.
505 mm
705 mm
675 mm
230 V 50/60 Hz 8000 w
8000 w
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products