missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Garcin™ Bactinyl Disinfectant Detergent
Use Garcin™ Bactinyl Disinfectant Detergent as a janitorial cleaner and disinfectant. Its active substances include peroxides, quaternary ammoniums and ethanol.
POA
Specifications
Product Type | Bactinyl Disinfectant Detergent |
---|---|
Form | Liquid |
pH | 10 ± 1 |
Appearance | Opalescent |
Certifications/Compliance | EN 1040, EN 1276, NF T 72-151 spectrum 5, NF T 72-190 NF T 72-171et, NF EN 1275 and NF EN 1650, NF T 72-180, EC 0398 |
Description
- Sterile
- Rinse not justified if thorough drying
- Odorless
- Inactivation of the HIV virus in 5 minutes
- Approved for food contact
Specifications
Bactinyl Disinfectant Detergent | |
10 ± 1 | |
EN 1040, EN 1276, NF T 72-151 spectrum 5, NF T 72-190 NF T 72-171et, NF EN 1275 and NF EN 1650, NF T 72-180, EC 0398 | |
1 Ea. |
Liquid | |
Opalescent | |
4°C |