missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Gas Distribution Tube, Type A

Gas Distribution Tube with centered bowl.

Brand:  ROBU VitraPOR™ 19251

Additional Details : Weight : 0.19000kg

 View more versions of this product

Product Code. 16812986

  • 178.00 € / Pack of 5
Estimated Shipment: 30-05-2024
to see stock.
Add to basket

Description

Description

  • For fine dispersion of gases in liquids
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
Type A
1
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers